missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Potassium tantalum isopropoxide, 5% w/v in isopropanol

Brand:  Alfa Aesar™ 039167.AC

Code : NEW

Additional Details : CAS Number : 21864-26-6 Weight : 0.01000kg Transport : UN number : 1219 Chem class : 3 Pack group : II

Quantity 25mL

 View more versions of this product

Product Code. 11383538

  • £54.91 / 25mL


Estimated Shipment date
from Supplier 21-06-2018
Add to basket

Specifications

Chemical Identifiers

Potassium tantalum isopropoxide
KTa[OCH(CH3)2]6
MFCD00210644
21864-26-6
574.57
UN1219

Specifications

Potassium tantalum isopropoxide
Liquid
574.57
UN1219
Colorless to pale yellow
25mL
Moisture Sensitive
21864-26-6
KTa[OCH(CH3)2]6
MFCD00210644
Alcohol-like
5% w/v in isopropanol
12° (54°F)
Videos
SDS