missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium superoxide, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
500 g
Packaging:
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 400°C |
Color | Yellow |
Physical Form | Chunks and/or Powder |
Quantity | 500 g |
Linear Formula | KO2 |
Solubility Information | Solubility in water: reacts |
Formula Weight | 71.1 |
Packaging | Plastic bottle |
Chemical Name or Material | Potassium superoxide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'