missing translation for 'onlineSavingsMsg'
Learn More

Potassium sulfate, Puratronic™, 99.997% (metals basis), Thermo Scientific Chemicals

Catalog Number p-7048551
33.06 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
7778-80-5
K2O4S
174.25
MFCD00011388
OTYBMLCTZGSZBG-UHFFFAOYSA-L
potassium sulfate, dipotassium sulfate, potassium sulphate, sulfuric acid dipotassium salt, sal polychrestum, sulfuric acid, potassium salt, arcanum duplicatum, kalium sulphuricum, caswell no. 702, sulfuric acid, dipotassium salt
24507
CHEBI:32036
dipotassium;sulfate
[K+].[K+].[O-]S([O-])(=O)=O
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
7778-80-5
K2O4S
174.25
MFCD00011388
OTYBMLCTZGSZBG-UHFFFAOYSA-L
potassium sulfate, dipotassium sulfate, potassium sulphate, sulfuric acid dipotassium salt, sal polychrestum, sulfuric acid, potassium salt, arcanum duplicatum, kalium sulphuricum, caswell no. 702, sulfuric acid, dipotassium salt
24507
CHEBI:32036
dipotassium;sulfate
[K+].[K+].[O-]S([O-])(=O)=O

Potassium sulfate principal use is as a fertilizer. It is also used occasionally in the production of glass and as a flash reducer in artillery propellant charges. It reduces muzzle flash, flareback and blast overpressure.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 1689°C
Melting Point 1,069°C
Color White
Physical Form Powder
Quantity 5 g
Linear Formula K2SO4
Merck Index 14,7674
Solubility Information Soluble in water and glycerol. Insoluble in alcohol,acetate and carbon disulfide.
Formula Weight 174.27
Percent Purity 99.997%
Grade Puratronic™
Odor Odorless
Density 2.662 g/mL
Assay (metals basis)
Chemical Name or Material Potassium sulfate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'