missing translation for 'onlineSavingsMsg'
Learn More

Potassium Standard For ICP, 1000μg/mL in 2% HNO3, Assurance Grade, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Brand:  SPEX Certiprep PLK2-2Y

Code : 04

Additional Details : CAS Number : 7757-79-1 Weight : 0.12600kg Transport : UN number : 3264 Chem class : 8 Pack group : III


Quantity 125mL
Packaging Triple-leached LDPE bottle

 View more versions of this product

Product Code. 10162822

  • £39.00 / 125mL


Estimated Shipment
18-06-2018
Add to basket


Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Description

Description

AnalyteConcentration
K1000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Chemical Identifiers

Potassium Standard
AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

Specifications

Potassium Standard
Assurance Grade
Assurance Certificate of Analysis reports up to 30 elements at ppb levels.
2% HNO3
AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
7732-18-5
Triple-leached LDPE bottle
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
125mL
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 1000

Signal Word
  • Warning

Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P264-Wash thoroughly after handling.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains¬†: nitric acid, potassium nitrate

Documentation

Product intended for laboratory use only