missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium sorbate, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
250 g
1 kg
5 kg
Packaging:
Plastic bottle
Plastic drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Acidity | Passes test |
Melting Point | 270.0°C |
Color | White to Cream |
Loss on Drying | 1% (105°C, 3 hrs) max. |
Infrared Spectrum | Authentic |
Assay Percent Range | 99% |
Packaging | Plastic drum |
Linear Formula | CH3CH=CHCH=CHCOOK |
Quantity | 5 kg |
Merck Index | 15, 8847 |
Show More |
Safety and Handling
Product Identifier
- Potassium sorbate
- Warning
- Serious eye damage/eye irritation Category 2
- H319-Causes serious eye irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P362-Take off contaminated clothing.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'