missing translation for 'onlineSavingsMsg'
Learn More

Potassium, solid, 99% (metals basis), Thermo Scientific Chemicals

Catalog Number 11321398
Click to view available options
Quantity:
1 g
5 g
7440-09-7
K
39.10
MFCD00133776
ZLMJMSJWJFRBEC-UHFFFAOYSA-N
kalium, potassium, metal, potassium, liquid alloy, unii-rwp5ga015d, rwp5ga015d, hsdb 698, monopotassium, potasio, mono-potassium, atom
5462222
CHEBI:26216
potassium
[K]
7440-09-7
K
39.10
MFCD00133776
ZLMJMSJWJFRBEC-UHFFFAOYSA-N
kalium, potassium, metal, potassium, liquid alloy, unii-rwp5ga015d, rwp5ga015d, hsdb 698, monopotassium, potasio, mono-potassium, atom
5462222
CHEBI:26216
potassium
[K]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Silver
Appearance Silver colored solid
Density 0.86g/cm3
Assay Percent Range 99.95%
Quantity 5 g
Odor Odorless
Chemical Name or Material Potassium

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'