Learn More
Potassium, solid, 99% (metals basis), Thermo Scientific Chemicals
Description
Potassium combines under pressure with silver metal to form 2 new compounds: K2Ag and K3Ag. This chemical is important in neuron (brain and nerve) function, and in influencing osmotic balance between cells and the interstitial fluid, with their distribution mediated in all animals (but not in all plants) by the so-called Na+/K+-ATPase pump. It is also used to prevent muscle contraction and the sending of all nerve impulses in animals through action potentials. Potassium ion is a nutrient necessary for human life and health. It is used as reducing agent in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
UN Number | UN2257 |
Quantity | 1 g |
Solubility Information | Soluble in water. |
Percent Purity | 99.95% |
Odor | Odorless |
Packaging | Breakseal Ampules |
Assay | (metals basis) |
Chemical Name or Material | Potassium |
RUO – Research Use Only