missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Sodium Tartrate Tetrahydrate, Certified AR for Analysis, Fisher Chemical™
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
Description
Potassium Sodium Tartrate Tetrahydrate, Certified AR for Analysis, Fisher Chemical, Quantity: 500g, CAS: 6381-59-5, White, Melting Point: 70 deg.C, Molecular Formula: C4H12KNaO10, Molecular Weight: 282.218, Packaging: Plastic powder jar, pH: 5.5 to 8.5
Specifications
Melting Point | 70°C |
Color | White |
pH | 5.5 to 8.5 |
Physical Form | Powder/Solid |
Quantity | 2.5 kg |
Formula Weight | 282.22g/mol |
Packaging | Plastic powder jar |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'