missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium pyrophosphate, anhydrous, 96% min, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 kg
5 kg
Description
Used in food industry. It is used for cyanide free electroplating, metal cleaning. It is used as a buffer, dye, dispersant and high capability detergent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Boiling Point | (decomposition) |
Melting Point | 1090°C |
Color | White |
Physical Form | Granules |
Quantity | 1 kg |
Linear Formula | K4P2O7 |
Merck Index | 14,7663 |
Solubility Information | Very soluble in water. Insoluble in ethyl alcohol. |
Formula Weight | 330.34 |
Percent Purity | ≥96% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'