missing translation for 'onlineSavingsMsg'
Learn More

Potassium Phosphate, Tribasic, n-Hydrate, Powder, BAKER ANALYZED™ Reagent, J.T. Baker™

Catalog Number p-5390856
427.50 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.


Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
This item is not returnable. View return policy
7778-53-2
K3O4P
212.265
LWIHDJKSTIGBAC-UHFFFAOYSA-K
potassium phosphate, tripotassium phosphate, potassium phosphate tribasic, phosphoric acid, tripotassium salt, potassium orthophosphate, tripotassium orthophosphate, caswell no. 700a, potassium phosphate, tribasic, potassium phosphate k3po4, ccris 7321
62657
tripotassium;phosphate
[O-]P(=O)([O-])[O-].[K+].[K+].[K+]
This item is not returnable. View return policy
7778-53-2
K3O4P
212.265
LWIHDJKSTIGBAC-UHFFFAOYSA-K
potassium phosphate, tripotassium phosphate, potassium phosphate tribasic, phosphoric acid, tripotassium salt, potassium orthophosphate, tripotassium orthophosphate, caswell no. 700a, potassium phosphate, tribasic, potassium phosphate k3po4, ccris 7321
62657
tripotassium;phosphate
[O-]P(=O)([O-])[O-].[K+].[K+].[K+]

High quality chemicals for laboratory and specialized industrial use

Physical Form Powder
Quantity 500 g
Grade BAKER ANALYZED™ Reagent
Packaging Poly Bottle
Chemical Name or Material Potassium Phosphate, Tribasic, n-Hydrate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'