missing translation for 'onlineSavingsMsg'
Learn More

Potassium Phosphate NF GenAR Crystal, Macron Fine Chemicals™

Catalog Number 15578434
140.60 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.


Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
POLYSTORMOR
Poly
This item is not returnable. View return policy
7778-77-0
H2KO4P
136.08
MFCD00011401 MFCD00147253
GNSKLFRGEWLPPA-UHFFFAOYSA-M
potassium dihydrogen phosphate, monopotassium phosphate, potassium phosphate monobasic, potassium phosphate, monobasic, potassium acid phosphate, phosphoric acid, monopotassium salt, monobasic potassium phosphate, monopotassium monophosphate, monopotassium dihydrogen phosphate, monopotassium orthophosphate
516951
CHEBI:63036
potassium dihydrogen phosphate
[K+].OP(O)([O-])=O
This item is not returnable. View return policy
7778-77-0
H2KO4P
136.08
MFCD00011401 MFCD00147253
GNSKLFRGEWLPPA-UHFFFAOYSA-M
potassium dihydrogen phosphate, monopotassium phosphate, potassium phosphate monobasic, potassium phosphate, monobasic, potassium acid phosphate, phosphoric acid, monopotassium salt, monobasic potassium phosphate, monopotassium monophosphate, monopotassium dihydrogen phosphate, monopotassium orthophosphate
516951
CHEBI:63036
potassium dihydrogen phosphate
[K+].OP(O)([O-])=O
Physical Form Crystalline
Quantity 500 g
Formula Weight 136.09
Grade NF
Packaging POLYSTORMOR
Chemical Name or Material Potassium Phosphate, Monobasic
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'