missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Phosphate, Monobasic, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
688.75 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.
Use promo code "23268" to get your promotional price.
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly bottle
Specifications
Physical Form | Crystals |
Quantity | 2.5 kg |
Formula Weight | 136.09 |
Grade | Meets ACS reagent chemical requirements |
Packaging | Poly bottle |
Specific Gravity | 1L = 2.34kg |
Chemical Name or Material | Potassium Phosphate, Monobasic |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'