missing translation for 'onlineSavingsMsg'
Learn More

Potassium Phosphate, Dibasic, Powder, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Catalog Number p-5390851
Click to view available options
Quantity:
2.5 kg
Packaging:
Poly Bottle
This item is not returnable. View return policy
7758-11-4
HK2O4P
174.17
ZPWVASYFFYYZEW-UHFFFAOYSA-L
dipotassium hydrogen phosphate, dipotassium phosphate, dipotassium hydrogenphosphate, dibasic potassium phosphate, potassium hydrogen phosphate, potassium phosphate, dibasic, potassium dibasic phosphate, potassium phosphate dibasic, phosphoric acid, dipotassium salt, dipotassium monophosphate
24450
CHEBI:32031
dipotassium;hydrogen phosphate
OP(=O)([O-])[O-].[K+].[K+]
This item is not returnable. View return policy
7758-11-4
HK2O4P
174.17
ZPWVASYFFYYZEW-UHFFFAOYSA-L
dipotassium hydrogen phosphate, dipotassium phosphate, dipotassium hydrogenphosphate, dibasic potassium phosphate, potassium hydrogen phosphate, potassium phosphate, dibasic, potassium dibasic phosphate, potassium phosphate dibasic, phosphoric acid, dipotassium salt, dipotassium monophosphate
24450
CHEBI:32031
dipotassium;hydrogen phosphate
OP(=O)([O-])[O-].[K+].[K+]

High quality chemicals for laboratory and specialized industrial use

Physical Form Powder
Quantity 2.5 kg
Formula Weight 174.18
Grade BAKER ANALYZED™ ACS Reagent
Packaging Poly Bottle
Specific Gravity 1L = 1.00kg
Chemical Name or Material Potassium Phosphate, Dibasic
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'