missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Phosphate, Dibasic, Endotoxin Tested, Multi-Compendial, U.S.P., J.T. Baker™
Click to view available options
Quantity:
500 g
2.5 kg
5 kg
Packaging:
Poly Bottle
Poly Pail
Specifications
Quantity | 5 kg |
Formula Weight | 174.18 |
Grade | Multi-Compendial/USP |
Packaging | Poly Pail |
Chemical Name or Material | Potassium Phosphate, Dibasic, Endotoxin Tested |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'