missing translation for 'onlineSavingsMsg'
Learn More

Potassium perrhenate, 99% (metals basis), Re 64%, Thermo Scientific Chemicals

Catalog Number p-7048755
Click to view available options
Quantity:
0.2 g
2 g
10 g
50 g
This item is not returnable. View return policy
10466-65-6
KO4Re
289.30
MFCD00011365
QFKRWIFGDGKWLM-UHFFFAOYSA-N
potassium perrhenate, potassiumperrhenate, k.o4re, potassium rhenium oxide kreo4, perrhenic acid hreo4 , potassium salt, potassium perrhenate vii, acmc-1bx96, perrhenic acid potassium salt, potassium oxido trioxo rhenium, potassium perrhenate kreo4
3661560
potassium;oxido(trioxo)rhenium
[K+].[O-][Re](=O)(=O)=O
This item is not returnable. View return policy
10466-65-6
KO4Re
289.30
MFCD00011365
QFKRWIFGDGKWLM-UHFFFAOYSA-N
potassium perrhenate, potassiumperrhenate, k.o4re, potassium rhenium oxide kreo4, perrhenic acid hreo4 , potassium salt, potassium perrhenate vii, acmc-1bx96, perrhenic acid potassium salt, potassium oxido trioxo rhenium, potassium perrhenate kreo4
3661560
potassium;oxido(trioxo)rhenium
[K+].[O-][Re](=O)(=O)=O

A powerful oxidizer

A powerful oxidizer. Also used for spectrometry, tungsten wire cathode coating color film and as catalyst. Potassium perrhenate is used as strong oxidizing agent and used in the synthesis of technetium and rhenium organometallic radiopharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 1360°C to 1370°C
Melting Point 550°C
Color White
Physical Form Crystalline
Quantity 0.2 g
UN Number UN1479
Solubility Information Soluble in water.
Formula Weight 289.3
Percent Purity 99%
Concentration or Composition (by Analyte or Components) Re 64%
Odor Odorless
Refractive Index 1.643
Density 4.887 g/mL
Assay (metals basis)
Chemical Name or Material Potassium perrhenate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'