missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Potassium peroxydisulfate, 99.99% (metals basis)

Brand:  Alfa Aesar™ 018227.14

Code : NEW

Additional Details : CAS Number : 7727-21-1 Weight : 0.02500kg Transport : UN number : 1492 Chem class : 1 Pack group : III

Quantity 25g

 View more versions of this product

Product Code. 11313437

  • £34.98 / 25g


Estimated Shipment date
from Supplier 24-05-2018
Add to basket

Specifications

Chemical Identifiers

Potassium peroxydisulfate
270.31
UN1492
7727-21-1
MFCD00011386
14,7656

Specifications

Potassium peroxydisulfate
Crystalline
(metals basis)
MFCD00011386
14,7656
Odorless
25g
7727-21-1
99.99%
270.31
UN1492
2.477g/mL
100°C (dec.)
Videos
SDS