missing translation for 'onlineSavingsMsg'
Learn More

Potassium Permanganate Solution, EU Pharmacopoeia Grade, Fisher Chemical™

Brand:  Fisher Chemical RP3621000

Code : 04

Additional Details : Weight : 1.00000kg Transport : UN number : 3082 Chem class : 1 Pack group : III


Product Code. 12943304

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Specifications

1000mL
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Potassium permanganate 0.200M (1N). Volumetric Solution (RP 362)

Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2

Hazard Statement
  • H411-Toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P273-Avoid release to the environment.
  • P391-Collect spillage.
  • P501-Dispose of contents/container to .