Potassium permanganate, Acros Organics


Overview
Brand: Acros Organics

Manufacturer Part Number: 447210025

UNSPSC: 12161500

Code: 29

Additional Details:
Additional Details: CAS Number: 7722-64-7 Weight: 2.55000kg Transport: UN number : 1490 Chem class : 1 Pack group : II


Quantity 2.5kg

Product Code. 13214809

Quantity Price
1 £ 165.6 / 2.50kg
EU Stock 8
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material Potassium permanganate
CAS 7722-64-7
Purity Grade ACS Reagent Grade
Molecular Formula KMnO4
Formula Weight 158.04

Specification

Formula Weight 158.04
Purity Grade ACS Reagent Grade
Quantity 2.5kg
CAS 7722-64-7
Molecular Formula KMnO4
Chemical Name or Material Potassium permanganate
Documentation

Technical documents