missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Potassium permanganate, 99+%, for analysis

Catalog Number p-3777008
38.00 GBP valid until 2024-03-30
Use promo code "21741" to get your promotional price.


28.60 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
Regulatory: This product contains a category 2 drug precursor according to Regulation (EC) No 273/2004, all customers must complete a declaration before delivery.
7722-64-7
KMnO4
MFCD00011364
VZJVWSHVAAUDKD-UHFFFAOYSA-N
potassium permanganate, chameleon mineral, condy's crystals, argucide, cairox, permanganate of potash, insta-perm, walko tablets, algae-k, solo san soo
516875
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
Regulatory: This product contains a category 2 drug precursor according to Regulation (EC) No 273/2004, all customers must complete a declaration before delivery.
7722-64-7
KMnO4
MFCD00011364
VZJVWSHVAAUDKD-UHFFFAOYSA-N
potassium permanganate, chameleon mineral, condy's crystals, argucide, cairox, permanganate of potash, insta-perm, walko tablets, algae-k, solo san soo
516875

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
TRUSTED_SUSTAINABILITY
Color Black-Green
Physical Form Crystalline Powder and/or Crystals
Quantity 500 g
Assay Percent Range 99+%
Linear Formula KMnO4
Fieser 01,942; 02,348; 04,412; 05,562; 08,416; 09,388; 10,330; 11,440; 12,413; 13,258; 14,267; 15,273; 16,283
Merck Index 15, 7776
SMILES [O-][Mn](=O)(=O)=O.[K+]
IUPAC Name potassium;permanganate
Molecular Weight (g/mol) 158.032
Formula Weight 158.04
Percent Purity ≥99.0%
Grade Analytical
Packaging Glass bottle
Chemical Name or Material Potassium permanganate, For analysis
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Potassium permanganate
Signal Word
  • Danger
Hazard Category
  • Oxidising solid Category 2
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 C
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity after repeated exposure Category 2
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H373-May cause damage to organs.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'