Potassium Permanganate, 0.1N Standardized Solution, Alfa Aesar™


Overview
Brand: Alfa Aesar

Manufacturer Part Number: 035590.K2

POTASSIUM PERMANGANATE .1NSTANDARD. SOLUTION 1L

UNSPSC: 12141724

Code: NEW

Additional Details:
Additional Details: CAS Number: 7722-64-7 Weight: 0.01000kg Transport: UN number : 1490 Chem class : 1 Pack group : II


Quantity 1L

Product Code. 11303697

Quantity Price
1 £ 20.56 / 1 L
Estimated Shipment date
from Supplier 20-02-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material Potassium permanganate
CAS 7722-64-7
Physical Form Liquid
Molecular Formula KMnO4
Formula Weight 158.03
MDL Number MFCD00011364

Specification

Formula Weight 158.03
Quantity 1L
Color Dark purple
Chemical Name or Material Potassium permanganate
EINECS Number 231-760-3
Physical Form Liquid
Molecular Formula KMnO4
Appearance Dark purple liquid
MDL Number MFCD00011364
CAS 7722-64-7