missing translation for 'onlineSavingsMsg'
Learn More

Potassium Permanganate, 0.1N Standardized Solution, Alfa Aesar™

Brand:  Alfa Aesar 035590.K7

UNSPSC : 12141724

Code : NEW

Additional Details : CAS Number : 7722-64-7 Weight : 0.01000kg

Quantity 4L

 View more versions of this product

Product Code. 11313697

  • £53.74 / 4 L


Estimated Shipment date
from Supplier 02-06-2017
Add to basket

Specifications Description & Specifications

Chemical Identifiers

Potassium permanganate
Liquid
158.03
7722-64-7
KMnO4
MFCD00011364

Specifications

Potassium permanganate
Liquid
Dark purple liquid
MFCD00011364
4L
7722-64-7
KMnO4
158.03
Dark purple
Documentation

Technical Documents