missing translation for 'onlineSavingsMsg'
Learn More

Potassium perchlorate, 99+%, for analysis, ACROS Organics™

Brand:  Acros Organics 221360025

Code : NEW

Additional Details : CAS Number : 7778-74-7 Weight : 2.50000kg Transport : UN number : 1489 Chem class : 5.1 Pack group : II

Packaging Plastic bottle
Quantity 2.5kg

 View more versions of this product

Product Code. 13438478

  • £227.48 / 2.50kg

Estimated Shipment
25-05-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Oxidation reagent
Specifications

Chemical Identifiers

7778-74-7
138.55
YLMGFJXSLBMXHK-UHFFFAOYSA-M
516900
[O-]Cl(=O)(=O)=O.[K+]
ClKO4
MFCD00011362
potassium perchlorate, perchloracap, astrumal, peroidin, perchloric acid, potassium salt, irenat, potassium hyperchloride, perchloric acid, potassium salt 1:1, potassium perchlorate kclo4, potassium perchlorate usp
potassium;perchlorate

Specifications

Potassium perchlorate
7778-74-7
ClKO4
MFCD00011362
potassium perchlorate, perchloracap, astrumal, peroidin, perchloric acid, potassium salt, irenat, potassium hyperchloride, perchloric acid, potassium salt 1:1, potassium perchlorate kclo4, potassium perchlorate usp
YLMGFJXSLBMXHK-UHFFFAOYSA-M
potassium;perchlorate
516900
Liquid
Analysis
30ppm max.
5ppm max.
Plastic bottle
Solubility in water: 17g/L (20°C). Other solubilities: 0.75 g/100mL water (0°C), 21.8 g/100mL water (100°C), practically insoluble in alcohol
Colorless to Yellow
2.5kg
p.a.
99+%
KClO4
15,7774
Solubility in water: 17g/L (20°C). Other solubilities: 0.75g/100mL water (0°C),21.8g/100mL water (100°C),practically insoluble in alcohol
[O-]Cl(=O)(=O)=O.[K+]
138.55
138.55
99+%
50ppm max.
30ppm max.
5ppm max.
200ppm max.
10ppm max.
400.0°C
Videos
SDS Documents