missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Alfa Aesar Potassium pentachlororhodate(III) hydrate, 99.9% (metals basis), Rh 29%

Brand:  Thermo Scientific Alfa Aesar 011053.06

Potassium pentachlororhodate(III) hydrate, 99.9% (metals basis), Rh 29%

Code : NEW

Additional Details : Weight : 0.00500kg

Product Code. 11321708

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.