missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Potassium oxide, super, 96.5%

Brand:  Alfa Aesar™ 014058.30

Code : NEW

Additional Details : CAS Number : 12030-88-5 Weight : 0.25000kg Transport : UN number : 2466 Chem class : 1 Pack group : I

Quantity 250g

 View more versions of this product

Product Code. 11317677

  • £94.20 / 250g


Estimated Shipment date
from Supplier 24-01-2018
Add to basket

Specifications

Chemical Identifiers

Potassium oxide
Super
MFCD00082411
Potassium superoxide
12030-88-5
71.1
UN2466

Specifications

Potassium oxide
12030-88-5
96.5%
MFCD00082411
Potassium superoxide
Odorless
Air & Moisture Sensitive
250g
Super
Powder
71.1
UN2466
2.14g/mL
2 to 10 Micron
≈400°C
Videos
SDS