missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Potassium oxide, super, 96.5%

Brand:  Alfa Aesar™ 014058.18

Code : NEW

Additional Details : CAS Number : 12030-88-5 Weight : 0.05000kg Transport : UN number : 2466 Chem class : 1 Pack group : I

Quantity 50g

 View more versions of this product

Product Code. 11327677

  • £25.20 / 50g


Estimated Shipment date
from Supplier 12-03-2018
Add to basket

Specifications

Chemical Identifiers

Potassium oxide
Super
MFCD00082411
Potassium superoxide
12030-88-5
71.1
UN2466

Specifications

Potassium oxide
12030-88-5
96.5%
MFCD00082411
Potassium superoxide
Odorless
Air & Moisture Sensitive
50g
Super
Powder
71.1
UN2466
2.14g/mL
2 to 10 Micron
≈400°C
Videos
SDS