missing translation for 'onlineSavingsMsg'
Learn More

Potassium nitrate, ReagentPlus™, Honeywell

Brand:  Honeywell Research Chemicals P8394-1KG

Code : NEW

Additional Details : CAS Number : 7757-79-1 Weight : 1.00000kg

Quantity 1kg

 View more versions of this product

Product Code. 15638060

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Chemical Identifiers

7757-79-1
101.102
FGIUAXJPYTZDNR-UHFFFAOYSA-N
CHEBI:63043
KNO3
MFCD00011409
24434

Specifications

Potassium nitrate
ReagentPlus™
KNO3
UN1486
[N+](=O)([O-])[O-].[K+]
101.102
CHEBI:63043
1kg
7757-79-1
≥99%
MFCD00011409
FGIUAXJPYTZDNR-UHFFFAOYSA-N
potassium;nitrate
24434
101.1g/mol
Videos
SDS