Potassium Nitrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

Overview
Brand: JT Baker

Manufacturer Part Number: 3190-01

500GR POTASSIUM NITRATE,CRYSTAL BAKER ANALYZED A.C.S. Reagent

UNSPSC: 12000000

Additional Details:
Additional Details: Weight: 0.92000kg


Packaging Wide Mouth Amber Glass Bottle
Quantity 500g

Product Code. 15538094

Quantity Price
1 £ 136.0 / 500g
Call for latest availability
+44 (0)1509 555 500
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material Potassium Nitrate
CAS 7757-79-1
Purity Grade Baker Analyzed™ ACS Reagent
Molecular Formula KNO3
Formula Weight 101.09

Specification

Purity Grade Baker Analyzed™ ACS Reagent
Physical Form Crystals
Formula Weight 101.09
Chemical Name or Material Potassium Nitrate
Packaging Wide Mouth Amber Glass Bottle
Specific Gravity 1L = 2.10kg
CAS 7757-79-1
Quantity 500g
Molecular Formula KNO3