missing translation for 'onlineSavingsMsg'
Learn More

Potassium nitrate, 99%, pure, ACROS Organics™

Brand:  Acros Organics 446370100

Code : 29

Additional Details : CAS Number : 7757-79-1 Weight : 10.05000kg Transport : UN number : 1486 Chem class : 5.1 Pack group : III

Packaging Plastic drum
Quantity 10kg

 View more versions of this product

Product Code. 12025390

  • £216.60 / 10kg


EU Stock 7
Add to basket

Sea freight only: For EU stock allow up to 15 days delivery.

Specifications

Chemical Identifiers

Potassium nitrate
Pure
KNO3
101.1
7757-79-1
99%
KNO3

Specifications

Potassium nitrate
Pure
99%
KNO3
Plastic drum
400.0°C
10kg
7757-79-1
99%
KNO3
101.1
Solubility in water: 320g/L (20°C). Other solubilities: soluble in glycerol, slightly soluble in alcohol
334.0°C
Videos
SDS