missing translation for 'onlineSavingsMsg'
Learn More

Potassium nitrate, 99%, pure, ACROS Organics™

Brand:  Acros Organics 446375000

Code : 29

Additional Details : CAS Number : 7757-79-1 Weight : 0.55000kg Transport : UN number : 1486 Chem class : 1 Pack group : III

Quantity 500g
Packaging Glass bottle

 View more versions of this product

Product Code. 12035390

  • £21.20 / 500g


Estimated Shipment
08-02-2018
Add to basket

Specifications

Chemical Identifiers

Potassium nitrate
Pure
KNO3
101.1
7757-79-1
99%
KNO3

Specifications

Potassium nitrate
Pure
99%
KNO3
Glass bottle
400.0°C
500g
7757-79-1
99%
KNO3
101.1
Solubility in water: 320g/L (20°C). Other solubilities: soluble in glycerol, slightly soluble in alcohol
334.0°C
Videos
SDS