missing translation for 'onlineSavingsMsg'
Learn More

Potassium nitrate, 99%, extra pure, ACROS Organics™

Brand:  Acros Organics 434430025

Code : 29

Additional Details : CAS Number : 7757-79-1 Weight : 2.55000kg Transport : UN number : 1486 Chem class : 1 Pack group : III

Quantity 2.5kg
Packaging Plastic Bottle

 View more versions of this product

Product Code. 10331750

  • £75.80 / 2.50kg


Estimated Shipment
19-03-2018
Add to basket

Specifications

Chemical Identifiers

Potassium nitrate
7757-79-1
99%
KNO3
101.1
Extra Pure
Extra Pure
99%
KNO3

Specifications

Potassium nitrate
7757-79-1
Fine Crystals or Crystalline Powder
99%
KNO3
Solubility in water: 320g/L (20°C). Other solubilities: soluble in glycerol, slightly soluble in alcohol
10ppm max.
0.001% max.
Solubility in water: 320g/L (20°C). Other solubilities: soluble in glycerol, slightly soluble in alcohol
400.0°C
334.0°C
Extra Pure
Extra Pure
99%
KNO3
101.1
0.002% max.
0.1% max.
Plastic Bottle
0.005% max.
White
2.5kg
Videos
SDS