missing translation for 'onlineSavingsMsg'
Learn More

Potassium nitrate, 99+%, for analysis, ACROS Organics™

Brand:  Acros Organics 205910100

Code : NEW

Additional Details : CAS Number : 7757-79-1 Weight : 10.00000kg Transport : UN number : 1486 Chem class : 5.1 Pack group : III


Packaging Plastic Drum
Quantity 10kg

 View more versions of this product

Product Code. 13426108

  • £160.00 / 10kg

Estimated Shipment
29-12-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

7757-79-1
101.1
FGIUAXJPYTZDNR-UHFFFAOYSA-N
24434
potassium;nitrate
KNO3
MFCD00011409
potassium nitrate, saltpeter, nitre, nitrate of potash, vicknite, nitric acid potassium salt, nitric acid, potassium salt, saltpetre, kaliumnitrat, kalii nitras
CHEBI:63043
[N+](=O)([O-])[O-].[K+]

Specifications

Potassium nitrate
99+%
KNO3
05,561
potassium nitrate, saltpeter, nitre, nitrate of potash, vicknite, nitric acid potassium salt, nitric acid, potassium salt, saltpetre, kaliumnitrat, kalii nitras
FGIUAXJPYTZDNR-UHFFFAOYSA-N
potassium;nitrate
24434
101.1
99+%
20ppm max
10ppm max.
10ppm max.
Plastic Drum
Solubility in water: 320g/L (20°C). Other solubilities: soluble in glycerol, slightly soluble in alcohol
400.0°C
334.0°C
10kg
7757-79-1
KNO3
MFCD00011409
15,7769
Solubility in water: 320g/L (20°C). Other solubilities: soluble in glycerol,slightly soluble in alcohol
[N+](=O)([O-])[O-].[K+]
101.1
CHEBI:63043
Fine Crystals
Analysis
20ppm max.
5ppm max.
10ppm max.
200ppm max.
50ppm max.
White
5.5 to 8 (5% soln. at 20°C)
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Potassium nitrate

Signal Word
  • Warning

Hazard Category
  • Oxidising solid Category 3

Hazard Statement
  • H272-May intensify fire; oxidiser.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P371+P380+P375-In case of major fire and large quantities: Evacuate area Fight fire remotely due to the risk of explosion.

SDS
Documents