missing translation for 'onlineSavingsMsg'
Learn More

Potassium nitrate, 99+%, ACS reagent, ACROS Organics™

Brand:  Acros Organics 424150025

Code : 29

Additional Details : CAS Number : 7757-79-1 Weight : 4.00000kg Transport : UN number : 1486 Chem class : 1 Pack group : III

Quantity 2.5kg
Packaging Glass bottle

 View more versions of this product

Product Code. 10754124

  • £84.10 / 2.50kg


EU Stock 6
Add to basket

Specifications

Chemical Identifiers

Potassium nitrate
7757-79-1
≥99%
KNO3
101.1
crystals
ACS Reagent Grade
99+%
KNO3
MFCD00011409

Specifications

Potassium nitrate
7757-79-1
Fine Crystals or Crystalline Powder
99+%
KNO3
MFCD00011409
05,561
50ppm max.
2.1
0.005% max.
3ppm max.
10ppm max.
5ppm max.
Solubility in water: 320g/L (20°C). Other solubilities: soluble in glycerol, slightly soluble in alcohol
400.0°C
334.0°C
2.5kg
crystals
ACS Reagent Grade
≥99%
KNO3
101.1
1486
15, 7769
20ppm max.
5ppm max.
5ppm max.
20ppm max.
Glass bottle
50ppm max.
30ppm max.
White
4.5 to 8.5 (5% soln. at 25°C)
Videos
SDS