missing translation for 'onlineSavingsMsg'
Learn More

Potassium nitrate, 99+%, Acros Organics

Brand:  Acros Organics 447820025

Code : NEW

Additional Details : CAS Number : 7757-79-1 Weight : 2.50000kg Transport : UN number : 1486 Chem class : 5.1 Pack group : III

Quantity 2.5kg
Packaging Plastic bottle

 View more versions of this product

Product Code. 13521620

  • £141.50 / 2.50kg


EU Stock 4
Add to basket

Specifications

Chemical Identifiers

Potassium nitrate
For biochemistry
KNO3
7757-79-1
99+%
101.1

Specifications

Potassium nitrate
For biochemistry
99+%
KNO3
Plastic bottle
400.0°C
2.5kg
7757-79-1
99+%
KNO3
101.1
Solubility in water: 320g/L (20°C). Other solubilities: soluble in glycerol, slightly soluble in alcohol
334.0°C
Videos