missing translation for 'onlineSavingsMsg'
Learn More

Potassium nitrate, 99.999%, (trace metal basis), ACROS Organics™

Brand:  Acros Organics 193800500

Code : 29

Additional Details : CAS Number : 7757-79-1 Weight : 0.10000kg Transport : UN number : 1486 Chem class : 1 Pack group : III

Quantity 50g
Packaging Glass bottle

 View more versions of this product

Product Code. 10114070

  • £89.90 / 50g


EU Stock 9
Add to basket

Specifications

Chemical Identifiers

Potassium nitrate
7757-79-1
99.999%
KNO3
MFCD00011409
99.999%
Trace Metal Basis
KNO3
101.1

Specifications

Potassium nitrate
7757-79-1
Crystals and Chunks
99.999%
KNO3
MFCD00011409
15, 7769
Glass bottle
0.001% max.
White
50g
99.999%
Trace Metal Basis
99.999%
KNO3
101.1
05,561
2.1
Solubility in water: 320g/L (20°C). Other solubilities: soluble in glycerol, slightly soluble in alcohol
400.0°C
334.0°C
Videos
SDS