missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium methoxide, pure, 0.1 N solution in Toluene/Methanol, Thermo Scientific Chemicals
Click to view available options
Quantity:
500 mL
2.5 L
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Colorless |
Linear Formula | CH3OK |
Concentration | 0.1N, exact strength on the certificate of analysis |
Density | 0.8500g/mL |
Solubility Information | Solubility in water: may decompose. |
Formula Weight | 70.13 |
Grade | Pure |
Packaging | Glass bottle |
Specific Gravity | 0.85 |
Quantity | 2.5 L |
Show More |
Safety and Handling
Product Identifier
- Potassium methoxide, 0.1N solution in toluene / methanol
- Danger
- Acute toxicity Category 3
- Acute toxicity Category 4
- Aspiration toxicity Category 1
- Flammable liquid Category 2
- Reproductive toxicity Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity after repeated exposure Category 2
- Specific target organ toxicity Category 3
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H361-Suspected of damaging fertility or the unborn child.
- H373-May cause damage to organs.
- H301-Toxic if swallowed.
- H331-Toxic if inhaled.
- H336-May cause drowsiness or dizziness.
- H332-Harmful if inhaled.
- H225-Highly flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P331-Do NOT induce vomiting.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'