missing translation for 'onlineSavingsMsg'
Learn More

Potassium methanesulfonate, 99%, Thermo Scientific Chemicals

Catalog Number p-7050805
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
2386-56-3
CH3KO3S
134.19
MFCD00070544
XWIJIXWOZCRYEL-UHFFFAOYSA-M
potassium methanesulfonate, methanesulfonic acid, potassium salt, potassium mesylate, potassium methylsulfonate, potassium methyl sulfonate, potassium methanesulphonate, unii-s99ib3em17, methanesulfonic acid potassium salt, methanesulfonic acid,potassium salt 1:1, methanesulfonic acid, potassium salt 1:1
23666501
potassium;methanesulfonate
CS(=O)(=O)[O-].[K+]
This item is not returnable. View return policy
2386-56-3
CH3KO3S
134.19
MFCD00070544
XWIJIXWOZCRYEL-UHFFFAOYSA-M
potassium methanesulfonate, methanesulfonic acid, potassium salt, potassium mesylate, potassium methylsulfonate, potassium methyl sulfonate, potassium methanesulphonate, unii-s99ib3em17, methanesulfonic acid potassium salt, methanesulfonic acid,potassium salt 1:1, methanesulfonic acid, potassium salt 1:1
23666501
potassium;methanesulfonate
CS(=O)(=O)[O-].[K+]

Used in pharmaceutical, chemical and biological research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color White
Odor Odorless
Linear Formula KCH3SO3
Quantity 25 g
Beilstein 3914899
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 134.19
Percent Purity 99%
Physical Form Powder or Lumps
Chemical Name or Material Potassium methanesulfonate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'