missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium metavanadate, 99.9% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
Description
Employed as corrosion inhibitors and anti-scaling agents. Used widely in chemical fertilizer industries, glass and ceramic industries, pharmaceutical industries. Also used in the process of pigment production for plastics, enamels, ceramics. Used for industrial gas desulphuration and H2S adsorption and elimination.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 520°C |
Physical Form | Powder |
Quantity | 25 g |
UN Number | UN2864 |
Solubility Information | Soluble in water. |
Formula Weight | 138.04 |
Percent Purity | 99.9% |
Odor | Odorless |
Assay | (metals basis) |
Chemical Name or Material | Potassium metavanadate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'