missing translation for 'onlineSavingsMsg'
Learn More

Potassium isopropoxide, 99% (metals basis), 5% w/v in isopropanol, Thermo Scientific Chemicals

Catalog Number p-7050633
Click to view available options
Quantity:
25 mL
100 mL
This item is not returnable. View return policy
6831-82-9
C3H7KO
98.186
MFCD00210641
WQKGAJDYBZOFSR-UHFFFAOYSA-N
potassium isopropoxide, potassium propan-2-olate, koipr, potassium isopropoxide w/v in isopropanol, potassium isopropoxide w/v in 2-propanol trace metals basis 25ml
23663646
potassium;propan-2-olate
CC(C)[O-].[K+]
This item is not returnable. View return policy
6831-82-9
C3H7KO
98.186
MFCD00210641
WQKGAJDYBZOFSR-UHFFFAOYSA-N
potassium isopropoxide, potassium propan-2-olate, koipr, potassium isopropoxide w/v in isopropanol, potassium isopropoxide w/v in 2-propanol trace metals basis 25ml
23663646
potassium;propan-2-olate
CC(C)[O-].[K+]

It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals. They are used in wide range of applications such as colorants and aroma chemicals. They are used in manufacturing detergents and biodiesel. They also act as catalysts in polymerization and isomerizations. Other uses include used for formation of ethers. Also acts as a moderately strong base for deprotonation and base catalysed reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless to Yellow
Flash Point 12°C (54°F)
Odor Alcohol-like
Linear Formula KOCH(CH3)2
Quantity 25 mL
UN Number UN3274
Sensitivity Air and moisture sensitive
Solubility Information It is soluble in isopropanol and some ethers. Insoluble in water.
Formula Weight 98.19
Percent Purity 99%
Concentration or Composition (by Analyte or Components) 5% w/v in isopropanol
Physical Form Liquid
Assay (metals basis)
Chemical Name or Material Potassium isopropoxide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'