missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium (iodomethyl)trifluoroborate, tech. 90%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
Organotrifluoroborate involved in synthesis of functionalized ethyltrifluoroborates, Suzuki-Miyaura cross-coupling reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Organotrifluoroborate involved in synthesis of functionalized ethyltrifluoroborates, Suzuki-Miyaura cross-coupling reactions.
Notes
Light Sensitive.Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, light.
Organotrifluoroborate involved in synthesis of functionalized ethyltrifluoroborates, Suzuki-Miyaura cross-coupling reactions.
Notes
Light Sensitive.Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, light.
Specifications
Melting Point | 242°C to 258°C |
Color | Brown to White |
Physical Form | Solid |
Quantity | 5 g |
Linear Formula | ICH2BF3K |
IUPAC Name | potassium trifluoro(iodomethyl)boranuide |
Formula Weight | 247.83 |
Percent Purity | 90% |
Grade | Technical |
Sensitivity | Light sensitive |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'