missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Iodide USP/FCC Powder, Macron Fine Chemicals™
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
POLYSTORMOR
Poly
Specifications
Physical Form | Powder |
Quantity | 2.5 kg |
Formula Weight | 166 |
Grade | FCC/USP |
Packaging | Poly |
Chemical Name or Material | Potassium Iodide |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'