missing translation for 'onlineSavingsMsg'
Learn More

Potassium iodide, ultra dry, 99.998% (metals basis), Thermo Scientific Chemicals

Catalog Number 11316938
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
7681-11-0
IK
166.003
MFCD00011405
NLKNQRATVPKPDG-UHFFFAOYSA-M
potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide
4875
CHEBI:8346
potassium;iodide
[K+].[I-]
This item is not returnable. View return policy
7681-11-0
IK
166.003
MFCD00011405
NLKNQRATVPKPDG-UHFFFAOYSA-M
potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide
4875
CHEBI:8346
potassium;iodide
[K+].[I-]

Potassium iodide is a simple iodide salt utilized as a fluorescence quenching agent. It is used in photography and as an analytical reagent, It is used in treating overactive thyroid and protecting the thyroid gland from the effects of radiation from inhaled or swallowed radioactive iodine. It is used as a nutrition supplement to prevent goitre and other iodine deficiency in human body.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 1330°C
Melting Point 681°C
Color White
Physical Form Beads
Quantity 25 g
Assay Percent Range 99.998% (metals basis)
Solubility Information Soluble in water (148g/100g at 25°C),alcohol (45.5mg/ml),methanol (125mg/ml),acetone (13.1mg/ml at 25°C),and ethanol (19.6mg/ml).
Formula Weight 166.01
Odor Odorless
Refractive Index 1.677
Sensitivity Hygroscopic
Appearance White beads
Vapor Pressure 1.33 hPa (1mm Hg) at 745°C
Density 3.12
Chemical Name or Material Potassium iodide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'