missing translation for 'onlineSavingsMsg'
Learn More

Potassium iodide, Free-flowing, ACS reagent, anhydrous, ≥99%, Honeywell Fluka™

Catalog Number 15636990
Click to view available options
Quantity:
100 g
500 g
1 kg
2.5 kg
Packaging:
Poly Bottle
This item is not returnable. View return policy
7681-11-0
IK
166.003
MFCD00011405
NLKNQRATVPKPDG-UHFFFAOYSA-M
potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide
4875
CHEBI:8346
potassium;iodide
[K+].[I-]
This item is not returnable. View return policy
7681-11-0
IK
166.003
MFCD00011405
NLKNQRATVPKPDG-UHFFFAOYSA-M
potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide
4875
CHEBI:8346
potassium;iodide
[K+].[I-]

Free-flowing, ACS reagent, anhydrous,≥99%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Boiling Point 1,323°C
Melting Point 681°C (literature)
CAS Max % 100.0000%
Physical Form Anhydrous
Quantity 500 g
Linear Formula KI
Formula Weight 166
Percent Purity >99%
Grade ACS Reagent
Packaging Poly Bottle
Density 3.12 g/cm3
Chemical Name or Material Potassium iodide, Free-flowing, ACS reagent, anhydrous, ≥99%
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'