Learn More
Potassium Hydroxide Solution 1M (1N), (Alcoholic), NIST Standard Solution Ready To Use, for Volumetric Analysis, Fisher Chemical™
Brand: Fisher Chemical J/6610/15
Code : 23
Additional Details : CAS Number : 1310-58-3 Weight : 1.00000kg Transport : UN number : 2924 Chem class : 3 Pack group : II
Quantity | 1L |
---|
Chemical Identifiers
3553 |
Specifications
3553 | |
1L | |
HDPE plastic bottle |
Colorless to Yellow | |
Liquid |