missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Hydroxide Solution 0.1M (0.1N), NIST Standard Concentrate, for Volumetric Analysis, Fisher Chemical™
Click to view available options
Quantity:
1 Ampule
6 Ampules
Specifications
pH | 13 |
Quantity | 6 Ampules |
IUPAC Name | potassium hydroxide |
Formula Weight | 56.11g/mol |
Packaging | Ampoule |
Physical Form | Liquid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'