missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hydroxide, Certified AR for Analysis, Pellets, meets analytical specification of Ph.Eur., BP, Fisher Chemical

Brand:  Fisher Chemical P/5640/60

16.93 GBP valid until 2020-06-30
Use promo code "13932" to get your promotional price.



Code : 22

Additional Details : CAS Number : 1310-58-3 Weight : 1.00000kg Transport : UN number : 1813 Chem class : 8 Pack group : II


Quantity 1kg

 View more versions of this product

Product Code. 10448990

Quantity Price
1 £43.97 / 1kg
4 £41.04 / 1kg
16 £38.99 / 1kg
In Stock 169

Please to purchase this item. Need a web account? Register with us today!



For a copy of the TSE/BSE statement please contact Technical Support on Tel: 01509 555888 or email our support team for further assistance
 

Specifications

Chemical Identifiers

1310-58-3
56.105
KWYUFKZDYYNOTN-UHFFFAOYSA-M
14797
potassium;hydroxide
HKO
3553
potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
CHEBI:32035
[OH-].[K+]

Specifications

1310-58-3
3553
KWYUFKZDYYNOTN-UHFFFAOYSA-M
potassium;hydroxide
14797
56.11g/mol
Plastic powder jar
360°C
White
HKO
potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
[OH-].[K+]
56.105
CHEBI:32035
Solid
13.5
1320°C
1kg
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Potassium hydroxide

Signal Word
  • Danger

Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 A

Hazard Statement
  • H290-May be corrosive to metals.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS Documents
View accessory products