missing translation for 'onlineSavingsMsg'
Learn More

Potassium hydroxide, Acculute Standard Volumetric Solution, Final Concentration 0.5N, Thermo Scientific Chemicals

Catalog Number p-7080443
Click to view available options
Quantity:
1 U
6 U
This item is not returnable. View return policy
1310-58-3
HKO
56.11
MFCD00003553
KWYUFKZDYYNOTN-UHFFFAOYSA-M
potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
14797
CHEBI:32035
potassium;hydroxide
[OH-].[K+]
This item is not returnable. View return policy
1310-58-3
HKO
56.11
MFCD00003553
KWYUFKZDYYNOTN-UHFFFAOYSA-M
potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
14797
CHEBI:32035
potassium;hydroxide
[OH-].[K+]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless
UN Number UN1814
Formula Weight 56.11
Quantity 1 U
Physical Form Liquid
Chemical Name or Material Potassium hydroxide Acculute Standard Volumetric Solution, Final Concentration 0.5N

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'