missing translation for 'onlineSavingsMsg'
Learn More

Potassium hydroxide, 99.98% (metals basis), 85% min, Thermo Scientific Chemicals

Catalog Number p-4408463
52.06 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
1310-58-3
HKO
56.11
MFCD00003553
KWYUFKZDYYNOTN-UHFFFAOYSA-M
potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
14797
CHEBI:32035
potassium;hydroxide
[OH-].[K+]
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
1310-58-3
HKO
56.11
MFCD00003553
KWYUFKZDYYNOTN-UHFFFAOYSA-M
potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
14797
CHEBI:32035
potassium;hydroxide
[OH-].[K+]

Potassium hydroxide acts as the precursor for most of the soft and liquid soaps, and for numerous potassium-containing chemicals. It is widely used in the manufacture of detergents, fertilizer, rayon, paper, dyestuffs, various drugs, drug intermediates, fine chemicals, and petroleum products. In the field of organic chemistry, potassium hydroxide acts as a base, and is used in the synthesis of several organic compounds. Further it finds application as an electrolyte in alkaline batteries, which are based on nickel-cadmium, nickel-hydrogen and manganese dioxide-zinc.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 1320°C to 1324°C
Melting Point 360°C
Color White
Physical Form Pellets
Quantity 100 g
Assay Percent Range 99.98% (metals basis)
Solubility Information Freely soluble in water. Soluble in ethanol.
Formula Weight 56.11
Odor Odorless
Density 2.044g/cm3 at 20°C
Chemical Name or Material Potassium hydroxide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'