missing translation for 'onlineSavingsMsg'
Learn More

Potassium hydroxide, 25% w/v aq. soln., Thermo Scientific Chemicals

Catalog Number 15462667
Click to view available options
Quantity:
500 mL
1 L
4 L
This item is not returnable. View return policy
1310-58-3
HKO
56.11
MFCD00003553
KWYUFKZDYYNOTN-UHFFFAOYSA-M
potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
14797
CHEBI:32035
[OH-].[K+]
This item is not returnable. View return policy
1310-58-3
HKO
56.11
MFCD00003553
KWYUFKZDYYNOTN-UHFFFAOYSA-M
potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
14797
CHEBI:32035
[OH-].[K+]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Physical Form Liquid
Quantity 500 mL
Linear Formula KOH
UN Number UN1814
IUPAC Name potassium;hydroxide
Formula Weight 56.11
Concentration or Composition (by Analyte or Components) 25% w/v aq. soln.
Chemical Name or Material Potassium hydroxide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'