Learn More
Potassium hydroxide, 1N solution in ethanol, AcroSeal™, Thermo Scientific Chemicals
Brand: Thermo Scientific Acros 429091000
10.53 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : 29
Additional Details : CAS Number : 64-17-5 Weight : 0.08600kg Transport : UN number : 2924 Chem class : 3 Pack group : II
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Potassium hydroxide | |
78.0°C | |
-117.0°C | |
KOH | |
0.950 to 1.050N | |
15, 7761 | |
Solubility in water: soluble. | |
KWYUFKZDYYNOTN-UHFFFAOYSA-M | |
potassium hydroxide | |
12°C | |
CHEBI:32035 | |
AcroSeal™ Glass Bottle | |
Liquid |
Colorless to Yellow | |
1310-58-3,64-17-5 | |
HKO | |
MFCD00003553 | |
0.8600g/mL | |
potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693 | |
100 mL | |
[OH-].[K+] | |
56.11 | |
14797 | |
56.11 | |
0.86 |
Safety and Handling
- Potassium hydroxide, 1N solution in ethanol
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 A
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H290-May be corrosive to metals.
- H225-Highly flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
For Manufacturing and Laboratory Use. Not for beverage use