missing translation for 'onlineSavingsMsg'
Learn More

Potassium hydroxide, 1.0N Standardized Solution in methanol, Thermo Scientific Chemicals

Catalog Number p-7058555
Click to view available options
Quantity:
1 L
4 L
20 L
1310-58-3
HKO
56.11
MFCD00003553
KWYUFKZDYYNOTN-UHFFFAOYSA-M
potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
14797
CHEBI:32035
potassium;hydroxide
[OH-].[K+]
1310-58-3
HKO
56.11
MFCD00003553
KWYUFKZDYYNOTN-UHFFFAOYSA-M
potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
14797
CHEBI:32035
potassium;hydroxide
[OH-].[K+]

Potassium hydroxide, 1.0N standardized solution in methanol is used for volumetric analysis in laboratories.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless
UN Number UN3286
Concentration 1.0N in methanol
Solubility Information Miscible with water.
Formula Weight 56.11
Odor Like alcohol
Quantity 1 L
Physical Form Liquid
Chemical Name or Material Potassium hydroxide Standardized Solution

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'